Spear sponges
Supplier: Harvard Apparatus
HARV59-8422EA 710 EUR
HARV59-8422
Spear sponges
Swabs Swabs for Clinical Use
These sterile cellulose sponges are designed for the removal of fluids during microsurgery.
  • Absorbs with only the tip immersed
  • Integral bendable plastic handle for added flexibility
  • Disposable for dissection use only
Suitable for spear applications.
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904