Cut resistant gloves, uvex C500 foam
Supplier: UVEX
6049409 6049410 6049411 6049407 6049408
112-4496EA 219 EUR
112-4496 112-4497 112-4498 112-4494 112-4495
Cut resistant gloves, uvex C500 foam
Gloves
These bamboo-rayon/Dyneema®/glass/polyamide gloves feature a HPE Soft Grip foam coating on palm and fingers. The Bamboo TwinFlex® technology ensures maximum cut resistance, exceptional abrasion resistance and outstanding wearing comfort. Suitable for multi-touch operations at industrial monitors.
  • Resist contact heat up to 100 °C
  • Excellent dexterity
  • Outstanding wearing comfort and tactile feel with innovative SoftGrip coating
  • Highest cut resistance level
  • Silicone-free according to imprint test
  • Moisture-proof
Typical applications include: Automotive industry, construction, brewery/beverage industry, glass industry, maintenance/servicing, metal industry, assembly works, paper industry, shipping/logistics, transportation works.
Colour: Lime/anthracite
EN 388 (4542)
Certifikace: EN 388:2016 (4X42C)
OEKO-Tex® Standard 100