Protective hand creams, Ligana® A Oleo-Tec
Supplier: Faweco Hautschutz
Ligana® Oleo-Tec
FAWE12983001EA 19.8 EUR
FAWE12983001
Protective hand creams, Ligana® A Oleo-Tec
Skincare Skin Creams Hand Creams
Ligana® Oleo-Tec Creme A are protective creams (o/w emulsion) against oil-insoluble contaminants such as oils, organic solvents, non-water soluble cooling lubricants, cutting oils, and other oily substances.
  • Absorbs quickly
  • Reduces skin irritations
  • Contains glycerine and beeswax
  • Silicone-free, perfumed
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904