Protective hand lotions, Ligana® Multi-tec
Supplier: Faweco Hautschutz
Ligana® Multi-tec
FAWE13042001EA 0 EUR
FAWE13042001
Protective hand lotions, Ligana® Multi-tec
Skincare Skin Creams Hand and Body Lotions
Ligana® Multi-tec is a skin protection lotion with dual effect, which is used when handling varying workplace substances. It does not leave any annoying greasy film on work pieces or tools. It is suitable for workplaces with constantly changing harmful substances, unspecific skin hazards or where hands are exposed to mechanical stress, e.g. from tools, fibreglass, wire wool or cardboard boxes, and also for workplaces which require the wearing of gloves.
  • Oil in water emulsion
  • Strengthens the upper layers of the skin and reduces skin irritation
  • Supports the natural regeneration capability of the skin
  • Contains glycerine, hamamelis and panthenol
  • Silicone-free, perfumed
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904