Surface disinfectant, Meliseptol® rapid
Supplier: B. Braun
18566 18564 18567 18565 18583 19108
BRAU18566EA 11.8 EUR
BRAU18566 BRAU18564 BRAU18567 BRAU18565 BRAU18583 BRAU19108
Surface disinfectant, Meliseptol® rapid
Cleaning Agents Disinfectants
Ready to use, alcoholic, surface disinfectant based on n-propanol and quaternary ammonium compound. It is active against bacteria, mycobacteria (M. terrae), yeasts and viruses (HBV, HIV, HCV, Adeno, Noro, Polio, Rota and Vaccinia).
  • Activity undiluted: 1 minute
  • Aldehyde- and alkylamine-free
  • Fully virucidal in 10 minutes
Informace pro objednávání:
1L spray bottle (BRAU18565): Sprayhead (148-4100) must be ordered separately.