Surface disinfectant, Meliseptol® Foam pure
Supplier: B. Braun
19288 19796
BRAU19288EA 70 EUR
BRAU19288 BRAU19796
Surface disinfectant, Meliseptol® Foam pure
Cleaning Agents Disinfectants
Ready to use formulation for surface disinfection of alcohol sensitive surfaces. e.g. ultrasound probes, space infusion pumps, Dialog+ Haemodialysis machines, artificial leather and more.
  • Based on n-propanol and quaternary ammonium compound
  • Active against bacteria, mycobacteria (M. terrae), fungi, viruses (HIV / HBV / HCV), Adeno- , Noro-, Polyoma-, Rota- and Vaccinia virus
  • Innovative aerosol-free foam spray head