Hand disinfectants, Sterillium® Virugard
Supplier: Bode
Sterillium® Virugard
115-1965EA 2.56 EUR
115-1965 BODE980012 BODL981075 115-1300 BODL981074 BODE980010 115-3990 BODL980339 BODL3066101
Hand disinfectants, Sterillium® Virugard
Skincare Skin Disinfectants Liquid and Foam Skin Disinfectants
Sterillium® Virugard is a ethanol based, highly effective virucidal hand disinfectant. Particularly suitable for all areas with an increased risk of infection.
  • Particularly active against enveloped and nonenveloped viruses
  • Bactericidal, yeasiticdal, fungicidal, tuberculocidal, mycobactericidal and virucidal
  • Recommended by the RKI for epidemic outbreaks including the noroviruses
  • Colourant- and fragrance-free
Upozornení: Use biocides safely. Always read the label and product information before use.
* These articles are only available in Germany.