Antiseptic spray, Cutasept® med F
Supplier: Bode
Cutasept® med F
148-4191EA 0 EUR
148-4191
Antiseptic spray, Cutasept® med F
First Aid Supplies First Aid Kit Components
Colourless antiseptic spray with ethanol, for use prior to injections, punctures and surgical procedures.
  • Ideal for use prior to blood glucose monitorings and insulin administration
  • Excellent skin compactibility, hypoallergenic
  • Effects: Bactericidal, yeasticidal, virucidal against eneveloped viruses (HBV, HIV, HCV)
  • Convenient invertible spray pump without propellants
Exposure time: 15 sec to 2 min