Cotton roving for pipette plugger
Supplier: Bellco Glass
BELC7750-22222EA 394 EUR
BELC7750-22222
Cotton roving for pipette plugger
Glass and Cotton Wool
Cotton roving made of 12 grain premium long fiber cotton.
Sufficient for approx. 35000 pipettes.
Balení: Coiled in plastic bag.
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904