ESD safety shoes, clogs, Classic, 31010, D222/D223
Supplier: Abeba
31010/36 31010/37 31010/38 31010/39 31010/40 31010/41 31010/42 31010/43 31010/44 31010/45 31010/46
ABEB31010/36EA 80.5 EUR
ABEB31010/36 ABEB31010/37 ABEB31010/38 ABEB31010/39 ABEB31010/40 ABEB31010/41 ABEB31010/42 ABEB31010/43 ABEB31010/44 ABEB31010/45 ABEB31010/46
ESD safety shoes, clogs, Classic, 31010, D222/D223
Shoes and Boots
ESD clog made from grained leather, non foldable, adjustable heel strap, and a PU sole (resistant to animal fats, oil and fuel).
  • Leather lining
  • Slip resistant PU sole
  • Protective steel toecap (200 J)
  • Adjustable instep strap with buckle fastening
For use in kitchens.
Colour: Black
Certifikace: EN 61340 (ESD); EN ISO 20345:2011; SB, A, E, FO, WRU, SRC