Disinfectant products, Korsolex® FF
Supplier: Hartmann Bode
BODL973889EA 0 EUR
BODL973889
Disinfectant products, Korsolex® FF
Korsolex ® FF is a broad spectrum and reliable acting, aldehydic instrument disinfectant that was developed especially for those users who want renounce the use of formaldehyde-containing products. Korsolex ® FF is preferably suitable for those areas where a short contact time with a broad spectrum of activity are required.
  • Formaldehyde-free
  • Excellent material compatibility
  • Compatible with chemo-thermal treatment
  • Bactericidal, fungicidal, tuberculocidal, mycobatericidal and virucidal against enveloped viruses