Surface disinfectants, concentrate, Mikrobac® basic

Supplier: Hartmann Bode
115-4023EA 0 EUR
BODL974845
Surface disinfectants, concentrate, Mikrobac® basic
Mikrobac® basic is suitable for the daily wipe disinfection of water resistant surfaces.

  • Contains benzyl-C12-18-alkyldimethylammonium chlorides 190 mg/g
  • Aldehyde-free, fragrance-free and colourant-free
  • Bactericidal, fungicidal and virucidal against enveloped viruses and rotavirus
  • Clear, slightly yellowish in appearance
  • pH value of 0,5% solution: approx 8,9
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR