Disinfectant product sprayers, Cutasept-G
Supplier: Hartmann
BODL975052EA 0 EUR
BODL975052 HART980367
Disinfectant product sprayers, Cutasept-G
Cleaning Agents Disinfectants
Cutasept® G is used in the surgical area, for pre-operative skin antisepsis with simultaneous marking of the disinfection area.
Contains propan-2-ol 63,0 g (72 vol.%), benzalkonium chloride 0,025 g with purified water and dyes sunset yellow S (E110), quinoline yellow (E104), brilliant black (E151) in 100 g of solution.
Bactericidal, fungicidal, tuberculocidal and virucidal against enveloped virus.
Areas of application: Prior to injections, catheterisation, punctures, blood sampling and small invasive procedures.
Upozornení: Not suitable for disinfection of large, open wounds and mucous membranes, not used on skin beneath tourniquet cuffs, avoid pooling and contact with eyes.