Hand cleanser dispenser, Eurospender, Basic
Supplier: Bode
Eurospender
BODL9770550EA 0 EUR
BODL9770550
Hand cleanser dispenser, Eurospender, Basic
Soap Dispensers and Lotion Soap Dispensers
The Eurospender Basic is designed for the use of hand desinfectants, detergents and body lotion.
  • Easy pump replacement in the front
  • Easy installation, easy operation, easy cleaning
  • Dosage amounts might be adjusted from 1 to 3 ml
Delivery information: Supplied with mounting device.