ESD glove liners, A198
Supplier: Portwest
A198GRRS A198GRRXS A198GRRXL A198GRRM A198GRRL
PRTWA198GRRSEA 16.4 EUR
PRTWA198GRRS PRTWA198GRRXS PRTWA198GRRXL PRTWA198GRRM PRTWA198GRRL
ESD glove liners, A198
Gloves
These liners feature a nylon and carbon fibre shell which diverts static electricity.
  • Fingertips with PU coating for precision applications
  • Excellent dexterity and durability
For use in electronics assembly, testing and precision work.
Colour: Grey/white
EN 388 (2132), EN 1149