Neutralisers for alkaline detergents, Dismoclean® 26 acid
Supplier: Hartmann Bode
BODL910980EA 0 EUR
BODL910980
Neutralisers for alkaline detergents, Dismoclean® 26 acid
Dismoclean ® 26 acid is a highly acidic pre-rinse or neutralisation solution based on phosphoric acid and complexing agents.
  • Formulation designed to reduce susceptibility to corrosion for most surgical steels
  • Application areas include instrument reprocessing for operating theatres, anaethesia, intensive care wards and microbiology laboratories