Slide cabinet
Supplier: Leica
LEIH3808500EEA 739 EUR
LEIH3808500E
Slide cabinet
Slide Holders
The slide cabinet is a storage system that can hold up to 6430 standard microscope slides.
  • Stackable
  • 14 drawers
  • Black base plinth or wheeled plinth
Informace pro objednávání: Other colours available in option.