Surface Cleaner, aerosol spray, Super-Degreaser
Supplier: King Tool
KING050502EA 13.9 EUR
KING050502
Surface Cleaner, aerosol spray, Super-Degreaser
Cleaning Agents Detergents and Surface Cleansers
Kitchen degreaser for removing fats and burnt on fat residues from ceramics, microwave interiors, glass oven doors, even barbecues.
  • Does not contain caustic soda
  • Compatible with metals, ceramics plastics and glass