Hand cleanser dispensers, Eurospender, 1 Plus Signal
Supplier: BODE CHEMIE
981251 981252 981253 981254 981256
BODE981251EA 80.3 EUR
BODE981251 BODE981252 BODE981253 BODE981254 BODE981256
Hand cleanser dispensers, Eurospender, 1 Plus Signal
Soap Dispensers and Lotion Soap Dispensers
These Eurodispenser 1 plus signal dispensers in high-visible colour are designed to improve hand-hygiene compliance. Colours draw users' attention and may improve compliance with hand disinfection.
  • Easy pump exchange through removal from the front, easy bottle replacement
  • Reliable robust metal dispenser, easy-to-use, not autoclavable
  • Adjustable dosing amount of 0,75 to 1,5 ml per actuation