Cartridge filters, Savana® PTFE
Supplier: Entegris
Savana®
514-0746EA 0 EUR
514-0746 514-0747 514-0748
Cartridge filters, Savana® PTFE
Filters Filter Cartridges
These cartridge filters are constructed of PTFE pleated membranes with PP supports to provide durability and cleanliness for filtration of process fluids used in the electronics industry.
  • Excellent chemical compatibility
  • Used for air filtration, stripper and IPA or solvent applications in flat panel display or hard disk industries
Membrane: Hydrophobic PTFE; Support, sleeve, caps and core: PP
Maximum operating temperature: 90 °C (for 70 mm Ø cartridge, in water); 80 °C (for 83 mm Ø cartridge, in water)
Maximum forward differential pressure: 4,0 bar at 25 °C
Maximum reverse differential pressure: 3,0 bar at 25 °C
Articles may not be available in all countries. Please contact your local Avantor office.