Culture tubes, glass, with screw cap
Supplier: Schuett-biotec
391-6136EA 349 EUR
391-6136
Culture tubes, glass, with screw cap
Tubes Culture Tubes
Borosilicate glass 3.3, with aluminium screw cap.
  • Wide neck, for easy inoculation and cleaning
  • Extra strong walls
  • Autoclavable/sterilizable
Ideal for cultivation, transporting and long-term storage.