Cannula filters, Erweka-style
Supplier: Quality Lab Accessories
QLAAFIL001-EW-1000EA 0 EUR
QLAAFIL004-EW-1000 QLAAFIL004-EW-100 QLAAFIL001-EW-100 QLAAFIL001-EW-1000
Cannula filters, Erweka-style
Filters Filter Tubes
Cannula filters are normally used with Erweka tablet dissolution baths for a large range of porosities.