Glove boxes, stainless steel combination, Protector®
Supplier: LABCONCO
5080172 5080170 5080165 5080167
LBCP5080172EA 122660 EUR
LBCP5080172 LBCP5080170 LBCP5080165 LBCP5080167
Glove boxes, stainless steel combination, Protector®
Glove Boxes and Glove Bags
These glove boxes with stainless steel liner provide inlet and outlet ULPA filtration and a leak-tight physical barrier to protect the operator from exposure to potentially hazardous materials and perform work with contamination-sensitive materials under vacuum or in an inert environment. Available with two or four glove ports (double version).
  • LCD display, large viewing area (double glove boxes have two viewing areas), space-saving inner and outer transfer chamber doors that pivot upward
  • Two internal valves to convert the glove box from a flow-through
More Product Information
Order Now
SPECIFICATIONS
Model Protector® (double) Protector®
Environmentally Preferable
Observation window (W×H) 2× 910×700 mm 910×700 mm
W×D×H ext. 2766×874×1161 mm 1613×874×1161 mm
Weight 590 kg 295 kg