Bolton-Hunter reagent (sulpho-SHPP)
Supplier: G Bioscienes
Synonyms:
Sulfo-SHPP
This sulpho-SHPP (water-soluble Bolton-Hunter Reagent) is designed to aid the labelling of proteins with radioactive iodine. Radioactive iodine (¹²⁵I) is routinely used by researchers to label proteins. The iodination of proteins can be performed enzymatically or chemically.
Bolton-Hunter Reagent is used to add tyrosine-like groups to end-terminal α-amino groups or ε-amino groups of lysine that result in increased protein iodination. The reaction is optimal at a pH of 8,5. This reagent is water soluble, and is ideal for proteins with masked or no tyrosine residues.
Formulation: Sulphosuccinimidyl-3-(4-hydroxyphenyl) propionate.
Formula:
C₁₃H₁₃NO₅ MW: 263,249 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00005515 CAS Number: 34071-95-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...