femtoCHROMO™- AP Substrate for chromogenic detection
Supplier: G Bioscienes
786-379 786-380 786-381 786-382 786-383
786-379EA 349 EUR
786-379 786-380 786-381 786-382 786-383
femtoCHROMO™- AP Substrate for chromogenic detection
Protein Linking and Labeling Systems
A ready to use modified BCIP (5-Bromo-4-Chloro-3’-Indolyphosphate p-Toluidine salt) and NBT (Nitro-Blue Tetrazolium Chloride) substrate that generates a black-purple insoluble precipitate in the presence of alkaline phosphatase.
  • Detects >5 ng
  • Ready to use, single detection step
  • High signal to background ratio and reproducibility
Delivery information: Supplied with an enhanced blocking agent, BLOT-QuickBlocker™, and a concentrated [10X] washing buffer, femtoTBST™ Buffer, to ensure low background staining. Optional AP labelled goat α-mouse or rabbit antibodies are also supplied.
Order Now
SPECIFICATIONS
Environmentally Preferable
Host
Learn more