Mineral oil, white (petroleum)
Supplier: G Bioscienes
RC-118EA 158 EUR
RC-118
Mineral oil, white (petroleum)
Mineral oil, white (petroleum)
Boiling Pt: 300 °C (1013 hPa)
Density: 0,872 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8042-47-5