Examination gloves, PVC, TT® Clear
Supplier: MEDICA EUROPE MAXXIM
480201 480203 480205 480207
MAXX480201EA 52.2 EUR
MAXX480201 MAXX480203 MAXX480205 MAXX480207
Examination gloves, PVC, TT® Clear
Gloves
These disposable PVC gloves contain no latex, NRL proteins or known accelerators. They are therefore suitable for people with a risk of allergy (type I and IV).
  • non sterile, smooth finish
  • Lightly powdered (corn starch)
  • Good touch sensitivity
  • High tensile strength
Colour: Natural
EN 455; AQL 1,5
Certifikace: EN 455; AQL 1,5