Waste disposal bags, Swirl®
Supplier: MELITTA HAUSHALTPRODUKTE
4008871215659 4008871215635
MELL4008871215659EA 9.9 EUR
MELL4008871215659 MELL4008871215635
Waste disposal bags, Swirl®
Bags Waste Disposal Bags
Elkay's water-soluble and eco-friendly bags are ideal for minimizing employee exposure to contaminated laundry.
  • Protects personnel from contamination and dissolves completely at a water temperature of 140 ° F or above
Informace pro objednávání: The binding tape is attached directly to the bag for a simple closure. Wet or damp laundry can safely be stored in the bag for several hours. Neither the laundry itself, nor the equipment nor the drainage system will be damaged if used correctly by the bags.