Silica gel, BAKER ANALYZED® ACS (6-18 Mesh) desiccant, with moisture indicator, J.T.Baker®
Supplier: Avantor
3401-01EA 0 EUR
3401-05 3401-01
Silica gel, BAKER ANALYZED® ACS (6-18 Mesh) desiccant, with moisture indicator, J.T.Baker®
Silica gel
6 to 18 mesh. Indicating (cobalt chloride). Lot analysis on label.
Color Change:
Blue.................Activated
Violet..........10% Moisture Absorbed
Pink............19% Moisture Absorbed
Pale Pink.......28% Moisture Absorbed
Formula: SiO₂
MW: 60,08 g/mol
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1710 °C
Density: 2,10 kg/m³
MDL Number: MFCD00011232
CAS Number: 63231-67-4
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Loss on Heating at 200°C ≤ 6.0 %
Suitability for Moisture Absorption ≥ 27 %
Suitability as Indicating Desiccant Passes Test
Mesh - Thru U.S. No. 6 Sieve ≥ 90 %
Mesh - Thru U.S. No. 18 Sieve ≤ 10 %
Color Change: Blue.................Activated Violet..........10% Moisture Absorbed Pink............19% Moisture Absorbed Pale Pink.......28% Moisture Absorbed