VWR®, Glass Beads for Grinding and Homogenisation
Supplier: VWR
VWR®
432-0064EA 317 EUR
432-0064
VWR®, Glass Beads for Grinding and Homogenisation
Beads Glass Beads
Small beads suitable for use with vortex mixers and tube homogenisers.
Molecular biology grade beads are treated to be free if DNase and RNase, making them suitable for a range of applications including disruption of mold and yeasts.