VWR® Ultra High Vacuum Aluminium Foil, All-Foils
Supplier: VWR Products
NPX11220118 NP112201.524
VWRMNPX11220118EA 625 EUR
VWRMNPX11220118 VWRMNP112201.524
VWR® Ultra High Vacuum Aluminium Foil, All-Foils
Aluminium Foils
Designed for use in controlled environments, ultra high vacuum aluminium foil is virtually free from contamination and residual rolling oils. Foil is available in several sizes and supplied on plastic cores. For extra protection against contamination, it is packaged in a plastic dispenser box and wrapped in film.