Molecular models
Supplier: Wiley
WILE3-527-32662-4EA 0 EUR
WILE3-527-32662-4
Molecular models
Models Molecular Biology Models
This set contains more than 240 atomic centers (annex C, H, O, N and P, S metals and halogens) and over 200 connections allow the assembly of even complex molecules from all fields of chemistry.
It has also 224 compounds in different lengths.
Informace pro objednávání: Includes a colour instruction booklet.