Bottle top vacuum filtration systems, MILLIPORE Express®
Supplier: Roth Carl
X338.1 X339.1 X340.1 X341.1 X342.1
ROTHX338.1EA 133 EUR
ROTHX338.1 ROTHX339.1 ROTHX340.1 ROTHX341.1 ROTHX342.1
Bottle top vacuum filtration systems, MILLIPORE Express®
Filters Bottle Top Filters
MILLIPORE Express® PLUS (PES)-membrane is a combination of bottle top filter and collecting bottle. For routine sterile filtration of tissue culture media, serum, additives and other aqueous solutions.
  • Low protein binding
  • High flow rates
  • Ideal for use under laminar flow