Peqlab, Strips of 8 PCR Tubes
Supplier: VWR Collection
732-2876EA 309 EUR
732-2876 PEQL82-0771-A 732-3229
Peqlab, Strips of 8 PCR Tubes
Tubes PCR Tubes
PP, natural or white.
  • Ultra-thin walls for rapid heat transfer
  • 'Universal fit' compatibility with a wide range of thermal cyclers
  • Domed or flat caps
  • Certified free from RNases, DNases and human DNA
  • Manufactured from virgin polypropylene under strict quality control guidelines
  • White strips and ultra-transparent tube caps ideal for use in 'Real Time' PCR
  • Low profile for reduced 'dead volume' and reduced sample condensation
Eight ultra-thin walled 0,15 (low profile) or 0,2 ml PCR tubes in a convenient strip for easier and quicker handling, ready for use on 96 format thermal cyclers. Strips are available with or without accompanying caps.