Peqlab, PCR Plates, 96-Well, Low Profile
Supplier: VWR Peqlab
732-3221EA 148 EUR
732-3221 732-3220
Peqlab, PCR Plates, 96-Well, Low Profile
Microplates PCR Plates
PP, non-skirted. Low profile plates significantly reduce the dead space between the film and the sample, which reduces the risk of sample condensation for enhanced reaction reproducibility.
  • Effective design with four overhanging edges for easy handling
  • Alphanumeric coding for quick sample identification
  • Certified free from RNase, DNase and human DNA
  • White plates are ideal for use in Real-Time PCR
  • Compatible with a wide range of thermal cyclers
  • Cut away corner (H12)