Peqlab, Adhesive Sealing Films
Supplier: VWR Collection
732-3212EA 167 EUR
732-3212 732-3218 732-3228
Peqlab, Adhesive Sealing Films
Seals Microplate Seals
Easy to use adhesive films for PCR and qPCR plates.
Adhesive PCR film
Transparent, self adhesive film. Temperature range: −20 to +120°C.
qPCR seal
Super-transparent, self adhesive film with minimum autofluorescence. Temperature range: −80 to +110°C.
Adhesive PCR foil seal
Metallised, self adhesive foil. Pierceable for sample recovery. Temperature range: −80 to +150°C.