Cleanroom gloves, nitrile, BioClean Nano 5™
Supplier: Ansell-Nitritex
BioClean Nano 5™
112-4304EA 291 EUR
112-4304 112-4308
Cleanroom gloves, nitrile, BioClean Nano 5™
Gloves
These nitrile gloves with beaded cuffs provide good resistance against a wide range of chemicals. They also have excellent ESD properties.
  • Contain no natural latex proteins, hypoallergenic
  • Extended leaching in pure deionised water ensures extremely low levels of chlorides and extractable metals
  • Textured fingers provide enhanced grip and dexterity
  • Anti-static
Suitable for use in Class 100, ISO 5 or M3.5 cleanrooms.
Certifikace: EN ISO 374-1 Type C, EN ISO 374-5 VIRUS, EN 421; Cat. III; AQL 1,5
Food contact approved.
Balení: 100 pieces per sealed inner PE bag; one sealed inner PE bag per sealed outer PE bag; 10 outer PE bags per carton liner; one carton liner per carton (1000 pieces).