Anti-ADORA2A Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-1456REA 671 EUR
BOSSBS-1456R
Anti-ADORA2A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ADORA2A (adenosine A2a receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR