Anti-PDI Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-1476REA 691 EUR
BOSSBS-1476R
Anti-PDI Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PDI
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR