Anti-Cathepsin D/CTSD Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-1615REA 717 EUR
BOSSBS-1615R
Anti-Cathepsin D/CTSD Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Cathepsin D/CTSD
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR