Anti-CXCR2 Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-1629REA 717 EUR
BOSSBS-1629R
Anti-CXCR2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CXCR2 (chemokine (C-X-C motif) receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR