Anti-HCLS1 Rabbit Polyclonal Antibody
Supplier: Aviva Systems Biology
ARP31928_P050 ARP31928_T100
AVIVARP31928_P050EA 401 EUR
AVIVARP31928_P050 AVIVARP31928_T100
Anti-HCLS1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HCLS1 (hematopoietic cell-specific Lyn substrate 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now