Anti-KLF3 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP32186_T100EA 335 EUR
AVIVARP32186_T100
Anti-KLF3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KLF3 (Kruppel-like factor 3 (basic))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR