Anti-ZUFSP Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP32274_P050EA 385 EUR
AVIVARP32274_P050
Anti-ZUFSP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZUFSP (zinc finger with UFM1-specific peptidase domain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR