Anti-POU1F1 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP32318_P050EA 385 EUR
AVIVARP32318_P050
Anti-POU1F1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: POU1F1 (POU class 1 homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR