Anti-NKX2-2 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP32338_P050EA 385 EUR
AVIVARP32338_P050
Anti-NKX2-2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NKX2-2 (NK2 homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR