Anti-NCOR2 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP32464_T100EA 335 EUR
AVIVARP32464_T100
Anti-NCOR2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NCOR2 (nuclear receptor corepressor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR