Anti-ACAT2 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP32791_T100EA 335 EUR
AVIVARP32791_T100
Anti-ACAT2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ACAT2 (acetyl-CoA acetyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR