Anti-ZFP36L1 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP33381_P050EA 385 EUR
AVIVARP33381_P050 AVIVARP33381_T100
Anti-ZFP36L1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZFP36L1 (ZFP36 ring finger protein-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR