Anti-MED27 Rabbit Polyclonal Antibody
Supplier: Aviva Systems Biology
AVIVARP33743_P050EA 385 EUR
AVIVARP33743_P050
Anti-MED27 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MED27 (mediator complex subunit 27)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat